Learn More
Medchemexpress LLC Cyt-1010 | 213769-33-6 | 99.7% | 624.73 g·mol⁻¹ | C35H40N6O5 | 5MG
SDP
Catalog No. 5000238478
Encompass_Preferred
CYT-1010 is a small-molecule mu-opioid receptor agonist described in patent WO2013173730A2 and supplied as a research reagent for in vitro and preclinical studies. It is provided as a solid powder and is intended for laboratory research use only, not for human or clinical applications.
  • Reported high potency with EC50s of 13.1 nM (β-arrestin recruitment) and 0.0053 nM (cAMP inhibition).
  • Molecular formula C35H40N6O5 and molecular weight 624.73 g·mol⁻¹.
  • Supplied as a solid powder with defined storage recommendations for powder and solution.
  • Available in multiple pack sizes, including small-scale milligram quantities for research workflows.
  • Includes supporting documentation such as certificate of analysis, safety data sheet, and analytical spectra.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-38478 Supplier Medchemexpress LLC Supplier No. HY1235345MG
CYT-1010 is a small-molecule mu-opioid receptor agonist described in patent WO2013173730A2 and supplied as a research reagent for in vitro and preclinical studies. It is provided as a solid powder and is intended for laboratory research use only, not for human or clinical applications.
  • Reported high potency with EC50s of 13.1 nM (β-arrestin recruitment) and 0.0053 nM (cAMP inhibition).
  • Molecular formula C35H40N6O5 and molecular weight 624.73 g·mol⁻¹.
  • Supplied as a solid powder with defined storage recommendations for powder and solution.
  • Available in multiple pack sizes, including small-scale milligram quantities for research workflows.
  • Includes supporting documentation such as certificate of analysis, safety data sheet, and analytical spectra.
  • For research use only; not for human or veterinary use.