Learn More
Enzo Life Sciences Cytochrome c monoclonal antibody (7H8.2C12) (100µg)
SDP
Supplier:  Enzo Life Sciences ADIAAM175E
Encompass_Preferred
Clone: 7H8.2C12. Host: Mouse. Isotype: IgG2b. Immunogen: Horse Cytochrome c. UniProt ID: P00004. Source: Purified from mouse ascites. Species reactivity: Human, Mouse, Rat, Sheep, Rabbit, Monkey, Hamster, Guinea pig, Bovine. Applications: IHC (PS), WB. Recommended Dilutions: Western Blot (1:1,000, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7143