Learn More
TARGETMOL CHEMICALS INC CYT-PTPEPSILON INHIBITOR 10MG
SDP
Catalog No. 502581615
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cyt-PTP(epsilon) Inhibitor-1 (A Inhibitor-1) is an inhibitor of cytosolic protein tyrosine phosphatase (epsilon). Cyt-PTP(epsilon) Inhibitor-1 blocks the dephosphorylation of c-Src and exhibits anti-osteoclastic activity. purity: 98%
Catalog No. 50-258-1615 Supplier TARGETMOL CHEMICALS INC Supplier No. T1092710MG
Also available in 1 mg 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cyt-PTP(epsilon) Inhibitor-1 (A Inhibitor-1) is an inhibitor of cytosolic protein tyrosine phosphatase (epsilon). Cyt-PTP(epsilon) Inhibitor-1 blocks the dephosphorylation of c-Src and exhibits anti-osteoclastic activity. purity: 98%