Learn More
Apexbio Technology LLC CYT387 1056634-68-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062589
Encompass_Preferred
CYT387 (CAS 1056634-68-4) is an aminopyrimidine derivative identified through high-throughput screening and optimized with structure-guided medicinal chemistry It functions as an inhibitor targeting Janus kinase family enzymes JAK1 JAK2 JAK3 and TYK2 with reported IC50 values of 11 nM 18 nM 155 nM and 17 nM respectively At concentrations between 500 1500 nM CYT387 selectively suppresses JAK2 signaling inhibits proliferation of JAK2-dependent hematopoietic cell lines and induces apoptosis with minimal effects on non-hematopoietic cells CYT387 is employed in experimental studies of JAK2-mediated signaling pathways and myeloproliferative neoplasms
Catalog No. 50-000-62589 Supplier Apexbio Technology LLC Supplier No. A41435.1
CYT387 (CAS 1056634-68-4) is an aminopyrimidine derivative identified through high-throughput screening and optimized with structure-guided medicinal chemistry It functions as an inhibitor targeting Janus kinase family enzymes JAK1 JAK2 JAK3 and TYK2 with reported IC50 values of 11 nM 18 nM 155 nM and 17 nM respectively At concentrations between 500 1500 nM CYT387 selectively suppresses JAK2 signaling inhibits proliferation of JAK2-dependent hematopoietic cell lines and induces apoptosis with minimal effects on non-hematopoietic cells CYT387 is employed in experimental studies of JAK2-mediated signaling pathways and myeloproliferative neoplasms