Learn More
Apexbio Technology LLC CYT387, 5mg. Cas: 1056634-68-4 MFCD: MFCD16038899. JAK-1/-2 inhibitor,ATP competitive
SDP
Supplier:  Apexbio Technology LLC A41435
Encompass_Preferred
CYT387, an aminopyrimidine derivative discovered by high-throughput enzyme and cell-based screening along with the optimization using structure-guided medicinal chemistry, is a potent and selective inhibitor of janus kinase 1 (JAK1), janus kinase 2 (JAK2) and tyrosine kinase 2 (TYK2). Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4891