Learn More
Enzo Life Sciences CYTAG CGH Labeling kit (100Reactions)
SDP
Supplier:  Enzo Life Sciences ENZ42671K100
Encompass_Preferred
Comparative genomic hybridization (aCGH) is a powerful diagnostic tool for detecting DNA copy number gains and losses associated with chromosome abnormalities. aCGH provides an understanding of genetic disorders, cancers and other genomic aberrations.   With optimized, proprietary reagents, the Enzo Life Sciences CGH Labeling Kits for oligo arrays produce high quality data using as little as 0.25 µg of genomic DNA, without a need for pre-amplification. Bulk quantity kit, 2 x 1000 reactions, is available upon special request. For the purification of labeled DNA, use PCR & Gel Clean-up columns, Prod No. ENZ-GEN100. Applications: CGH.
Catalog No. 50-201-2532
Add to Cart
Request a Quote