Learn More
Cayman Chemical Cytarabine 5′-monophosphate, Purity: ≥95%, CAS: 7075-11-8, 50 mg
SDP
Catalog No. 502911574
Encompass_Preferred
An active metabolite of cytarabine; is incorporated into DNA by DNA polymerase α, which slows the rate of DNA synthesis by approximately 2000-fold in a cell-free assay; inhibits nuclear and mitochondrial DNA replication in S. cerevisiae at 15 mM; improves survival in an L1210 murine leukemia model at 3.5-75.1 mg/kg
Catalog No. 50-291-1574 Supplier Cayman Chemical Supplier No. 3649650 MG
An active metabolite of cytarabine; is incorporated into DNA by DNA polymerase α, which slows the rate of DNA synthesis by approximately 2000-fold in a cell-free assay; inhibits nuclear and mitochondrial DNA replication in S. cerevisiae at 15 mM; improves survival in an L1210 murine leukemia model at 3.5-75.1 mg/kg