Learn More
Medchemexpress LLC HY-13605 1g , Cytarabine Cytosine β-D-arabinofuranoside;Cytosine Arabinoside;Ara-C CAS:147-94-4 Purity:98%
SDP
Catalog No. 501492284
Encompass_Preferred
Medchemexpress, HY-13605 1g Cytarabine Cytosine β-D-arabinofuranoside;Cytosine Arabinoside;Ara-C CAS:147-94-4 Formula:C9H13N3O5 IC50: 16 nM (DNA synthesis) Purity:98% Cytarabine is an antimetabolic agent and DNA synthesis inhibitor with IC 50 of 16 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2284 Supplier Medchemexpress LLC Supplier No. HY136051G
Medchemexpress, HY-13605 1g Cytarabine Cytosine β-D-arabinofuranoside;Cytosine Arabinoside;Ara-C CAS:147-94-4 Formula:C9H13N3O5 IC50: 16 nM (DNA synthesis) Purity:98% Cytarabine is an antimetabolic agent and DNA synthesis inhibitor with IC 50 of 16 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.