Learn More
Selleck Chemical LLC Cytarabine S1648-200mg
SDP
Catalog No. 503108490
Encompass_Preferred
Cytarabine is an antimetabolic agent and DNA synthesis inhibitor with IC50 of 16 nM in wild-type CCRF-CEM cells Cytarabine induces autophagy and apoptosis
Catalog No. 50-310-8490 Supplier Selleck Chemical LLC Supplier No. S1648200MG
Cytarabine is an antimetabolic agent and DNA synthesis inhibitor with IC50 of 16 nM in wild-type CCRF-CEM cells Cytarabine induces autophagy and apoptosis