Learn More
Medchemexpress LLC HY-13605 500mg Medchemexpress, Cytarabine CAS:147-94-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13605500MG
Encompass_Preferred
Medchemexpress, HY-13605 500mg Cytarabine CAS:147-94-4 Cytarabine, a nucleoside analog, causes S phase cell cycle arrest and inhibits DNA polymerase. Cytarabine inhibits DNA synthesis with an IC50 of 16 nM. Cytarabine has antiviral effects against HSV. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9008