Learn More
Selleck Chemical LLC Cytarabine 50mg 147-94-4 Cytarabin, Ara-C, Arabinofuranosyl Cytidine
SDP
Catalog No. NC0756755
Encompass_Preferred
Cytarabine (Cytarabin, Ara-C, Arabinofuranosyl Cytidine) is an antimetabolic agent and DNA synthesis inhibitor with IC50 of 16 nM in wild-type CCRF-CEM cells. Cytarabine induces autophagy and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0756755 Supplier Selleck Chemical LLC Supplier No. S164850MG
Cytarabine (Cytarabin, Ara-C, Arabinofuranosyl Cytidine) is an antimetabolic agent and DNA synthesis inhibitor with IC50 of 16 nM in wild-type CCRF-CEM cells. Cytarabine induces autophagy and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.