Learn More
Selleck Chemical LLC Cytarabine hydrochloride
SDP
Catalog No. 5000003817
Encompass_Preferred
Cytarabine is a pyrimidine nucleoside analog that is used mainly in the treatment of leukemia It is an antimetabolite antineoplastic agent that inhibits the synthesis of DNA
Catalog No. 50-000-03817 Supplier Selleck Chemical LLC Supplier No. S558225MG
Cytarabine is a pyrimidine nucleoside analog that is used mainly in the treatment of leukemia It is an antimetabolite antineoplastic agent that inhibits the synthesis of DNA