Learn More
TARGETMOL CHEMICALS INC CYTARABINE HYDROCHLORIDE 50MG
SDP
Catalog No. 502581427
Encompass_Preferred
Also available in 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cytarabine hydrochloride (Ara-C hydrochloride) is a nucleoside analog that causes S phase cell cycle arrest and inhibits DNA polymerase. Cytarabine inhibits DNA synthesis with an IC 50 of 16 nM. Cytarabine hydrochloride has antiviral effects against HSV. purity: 99%
Catalog No. 50-258-1427 Supplier TARGETMOL CHEMICALS INC Supplier No. T1272L50MG
Also available in 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cytarabine hydrochloride (Ara-C hydrochloride) is a nucleoside analog that causes S phase cell cycle arrest and inhibits DNA polymerase. Cytarabine inhibits DNA synthesis with an IC 50 of 16 nM. Cytarabine hydrochloride has antiviral effects against HSV. purity: 99%