Learn More
Medchemexpress LLC Cytarabine triphosph 10mg | 1179343-17-9 | 549.10 | C9H13N3Na3O14P3 | 10 MG

Supplier: Medchemexpress LLC HY115740A10MG
Cytarabine triphosphate trisodium is the trisodium salt of the active intracellular metabolite of cytarabine, provided as an aqueous solution for biochemical and cell biology research. It acts as a competitive inhibitor of DNA synthesis and is used to study nucleoside metabolism and antimetabolite mechanisms.
- Active intracellular metabolite of cytarabine (Ara-CTP).
- Competitive inhibitor of DNA synthesis.
- Supplied as an aqueous solution at 100 mM; common pack sizes include 5 mg, 10 mg, 25 mg, and 50 mg.
- Purity: 96.78%.
- Appearance: colorless to light yellow liquid.
- Salt form: trisodium; molecular weight 549.10; formula C9H13N3Na3O14P3.
- Storage: store at -20°C for long-term; stock solutions at -80°C (up to 6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.