Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cytarabine triphosph 25mg | 1179343-17-9 | 25 MG
SDP

Catalog No. 5000229228
Encompass_Preferred

Cytarabine triphosphate trisodium is the trisodium salt of cytarabine triphosphate and is used as a research reagent to study DNA synthesis inhibition and nucleoside metabolism.

  • Trisodium salt form supplied as a solution.
  • Pack size 25 mg, prepared as 100 mM x 500 μL in water for the 25 mg vial.
  • Molecular formula C9H13N3Na3O14P3 and molecular weight 549.10 g/mol.
  • Purity approximately 96.8%.
  • Storage: solution at -20°C; prepared stock solution stability: -80°C for 6 months, -20°C for 1 month.
  • Used as the active metabolite Ara-CTP in biochemical and cell-based assays targeting DNA synthesis.

Catalog No. 50-002-29228 Supplier Medchemexpress LLC Supplier No. HY115740A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cytarabine triphosphate trisodium is the trisodium salt of cytarabine triphosphate and is used as a research reagent to study DNA synthesis inhibition and nucleoside metabolism.

  • Trisodium salt form supplied as a solution.
  • Pack size 25 mg, prepared as 100 mM x 500 μL in water for the 25 mg vial.
  • Molecular formula C9H13N3Na3O14P3 and molecular weight 549.10 g/mol.
  • Purity approximately 96.8%.
  • Storage: solution at -20°C; prepared stock solution stability: -80°C for 6 months, -20°C for 1 month.
  • Used as the active metabolite Ara-CTP in biochemical and cell-based assays targeting DNA synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.