Learn More
Medchemexpress LLC Cytarabine triphosph 50mg | 1179343-17-9 | 549.10 | C9H13N3Na3O14P3 | 50 MG
SDP
Catalog No. 5000229227
Encompass_Preferred
Cytarabine triphosphate trisodium is the active intracellular triphosphate metabolite of cytarabine that competitively inhibits DNA synthesis. Supplied as an aqueous solution, it is intended for biochemical and cellular research requiring the active nucleotide form.
  • Active triphosphate metabolite of cytarabine.
  • Competitive inhibitor of DNA synthesis.
  • Supplied as an aqueous liquid for direct use or dilution.
  • Packaged as 50 mg (100 mM, 1 mL) solution.
  • Purity about 96.8% and recommended storage at -20°C.
Catalog No. 50-002-29227 Supplier Medchemexpress LLC Supplier No. HY115740A50MG
Cytarabine triphosphate trisodium is the active intracellular triphosphate metabolite of cytarabine that competitively inhibits DNA synthesis. Supplied as an aqueous solution, it is intended for biochemical and cellular research requiring the active nucleotide form.
  • Active triphosphate metabolite of cytarabine.
  • Competitive inhibitor of DNA synthesis.
  • Supplied as an aqueous liquid for direct use or dilution.
  • Packaged as 50 mg (100 mM, 1 mL) solution.
  • Purity about 96.8% and recommended storage at -20°C.