Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cytarabine triphosph 5mg | 1179343-17-9 | 549.10 g/mol | C9H13N3Na3O14P3 | 5 MG
SDP

Catalog No. 5000308118
Encompass_Preferred

Cytarabine triphosphate trisodium is the trisodium salt of cytarabine's active triphosphate metabolite, provided as a concentrated aqueous solution for biochemical and molecular biology research focused on DNA synthesis and nucleoside metabolism.

  • Supplied as a 5 mg solution at 100 mM (100 μL) in water.
  • Active metabolite that inhibits DNA synthesis, suitable for assay and mechanistic studies.
  • Liquid formulation, colorless to light yellow, for direct use in experiments.
  • Store at -20°C; solution stability reported up to 2 years.
  • Chemical formula C9H13N3Na3O14P3 and molecular weight approximately 549.1 g/mol.

Catalog No. 50-003-08118 Supplier Medchemexpress LLC Supplier No. HY115740A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cytarabine triphosphate trisodium is the trisodium salt of cytarabine's active triphosphate metabolite, provided as a concentrated aqueous solution for biochemical and molecular biology research focused on DNA synthesis and nucleoside metabolism.

  • Supplied as a 5 mg solution at 100 mM (100 μL) in water.
  • Active metabolite that inhibits DNA synthesis, suitable for assay and mechanistic studies.
  • Liquid formulation, colorless to light yellow, for direct use in experiments.
  • Store at -20°C; solution stability reported up to 2 years.
  • Chemical formula C9H13N3Na3O14P3 and molecular weight approximately 549.1 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.