Learn More
Aviva Systems Biology Corp CYTB Antibody - N-terminal region (ARP50256_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50256_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYTB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CYTB. Alternate names: MTCYBApplicationWB. Species reactivity: Human, Mouse, Rat
Catalog No. 50-014-53625