Learn More
Cayman Chemical Cytidine-d2, Purity: ≥99% deuterated forms (d1-d2), CAS: 40632-25-5, 1 mg
SDP
Catalog No. 502912958
Encompass_Preferred
An internal standard for the quantification of cytidine by GC- or LC-MS
Catalog No. 50-291-2958 Supplier Cayman Chemical Supplier No. 363201 MG
An internal standard for the quantification of cytidine by GC- or LC-MS