Learn More
Cytek Biosciences Inc CYTEK CFLUOR TBMNK KIT 8 COLOR
Supplier:  Cytek Biosciences Inc R740001
Encompass_Preferred
  • Optimized for rapid immunophenotyping workflows.
  • Enables comprehensive analysis of human peripheral blood mononuclear cells (PBMCs) and whole blood.
  • Facilitates precise identification of immune cell populations.
  • Includes 8 distinct colors for enhanced resolution.
  • Targets key immune cell types such as T cells, B cells, and various monocyte subsets.
  • Provides separate tubes with cFluor conjugated antibodies.
  • Standard test dilution of 5 µL per test.
  • Storage conditions of 2-8 °C, protected from light.
  • Offers 25 tests per kit.
  • Research use only, not for diagnostic purposes.
Catalog No. NC2590011