Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytek Biosciences Inc CYTEK FSP COMPBEADS

Catalog No. NC2450010
Encompass_Preferred

  • Mimics cell fluorescence using fluorescent conjugated antibodies.
  • Matches lymphocyte scatter properties for realistic compensation.
  • Broad immunoglobulin capture, including mouse, rat, and hamster IgG and IgM.
  • Enhances data accuracy by minimizing spectral overlap between fluorescence channels.
  • Simplifies workflow with a ready-to-use solution for compensation.
  • Provides optimal reference control for distinct positive and negative fluorescence populations.
  • Tests per kit: 100
  • Volume per test: 1 drop
  • Application: Flow cytometry
  • Storage: 2-8°C
  • Protection from light required.
  • Do not freeze.

Catalog No. NC2450010 Supplier Cytek Biosciences Inc Supplier No. B710011
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Mimics cell fluorescence using fluorescent conjugated antibodies.
  • Matches lymphocyte scatter properties for realistic compensation.
  • Broad immunoglobulin capture, including mouse, rat, and hamster IgG and IgM.
  • Enhances data accuracy by minimizing spectral overlap between fluorescence channels.
  • Simplifies workflow with a ready-to-use solution for compensation.
  • Provides optimal reference control for distinct positive and negative fluorescence populations.
  • Tests per kit: 100
  • Volume per test: 1 drop
  • Application: Flow cytometry
  • Storage: 2-8°C
  • Protection from light required.
  • Do not freeze.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.