Learn More
Rarecyte ArgoFluor Mounting Media, 10mL
Catalog No. NC1406505
Encompass
Solution for adhering a coverslip to a microscope slide.  To be used with RareCyte’s AccuCyte/RarePlex liquid biopsy products and assays.  DO NOT USE with RareCyte’s Orion/ArgoFluor products.
Catalog No. NC1406505 Supplier Rarecyte Supplier No. 421056000
Solution for adhering a coverslip to a microscope slide.  To be used with RareCyte’s AccuCyte/RarePlex liquid biopsy products and assays.  DO NOT USE with RareCyte’s Orion/ArgoFluor products.