Learn More
Cayman Chemical CytIdIn 3-monphosphate 50mg
SDP
Catalog No. 502946508
Encompass_Preferred
A ribonucleotide; produced by hydrolysis of cytidine 2’,3’-cyclic monophosphate via RNase; inhibits RNase; can be dephosphorylated to cytidine by 3’-nucleotidase
Catalog No. 50-294-6508 Supplier Cayman Chemical Supplier No. 3383350 MG
A ribonucleotide; produced by hydrolysis of cytidine 2’,3’-cyclic monophosphate via RNase; inhibits RNase; can be dephosphorylated to cytidine by 3’-nucleotidase