Learn More
Cayman Chemical Cytidine 5'-methylenediphosphate (sodium salt), Purity: ≥95%, 2 mg
SDP
Catalog No. 502916766
Encompass_Preferred
An inhibitor of ecto-5’-nucleotidase (Ki = 898 nM)
Catalog No. 50-291-6766 Supplier Cayman Chemical Supplier No. 383722 MG
An inhibitor of ecto-5’-nucleotidase (Ki = 898 nM)