Learn More
Cayman Chemical Cytidine 5'-monophosphate, Purity: ≥95%, CAS: 63-37-6, 1 g
SDP
Catalog No. 502912776
Encompass_Preferred
A pyrimidine nucleotide; used in the synthesis of RNA; an intermediate in the synthesis of phospholipids
Catalog No. 50-291-2776 Supplier Cayman Chemical Supplier No. 351161 G
A pyrimidine nucleotide; used in the synthesis of RNA; an intermediate in the synthesis of phospholipids