Learn More
Cayman Chemical Cytidine 5'-triphosphate (sodium salt hydrate), Purity: ≥95%, CAS: 652154-13-7, 100 mg
SDP
Catalog No. 502966682
Encompass_Preferred
A pyrimidine nucleoside triphosphate that is involved in a variety of biochemical reactions, including RNA synthesis, the formation of PC, and sialylation
Catalog No. 50-296-6682 Supplier Cayman Chemical Supplier No. 18147100 MG
A pyrimidine nucleoside triphosphate that is involved in a variety of biochemical reactions, including RNA synthesis, the formation of PC, and sialylation