Learn More
Cayman Chemical CytIdIn5O 2thIodIphosph 2.1mg
SDP
Catalog No. 502918736
Encompass_Preferred
A non-hydrolyzable derivative of CDP with antioxidant activity; scavenges free radicals in a TEAC assay from 5-20 µM
Catalog No. 50-291-8736 Supplier Cayman Chemical Supplier No. 385602.1 MG
A non-hydrolyzable derivative of CDP with antioxidant activity; scavenges free radicals in a TEAC assay from 5-20 µM