Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC α-cytidine | 13913-16-1 | 98.7% | 243.22 g/mol | C9H13N3O5 | 5 MG
SDP

Catalog No. 5000283877
Encompass_Preferred

α-Cytidine is a purine nucleoside analog used in biochemical and pharmacological research to investigate antitumor activity and nucleoside metabolism. It inhibits DNA synthesis and can induce apoptosis, making it relevant for studies of lymphoid malignancies and mechanistic assays.

  • Purity approximately 98.7% for reliable experimental results.
  • Molecular weight 243.22 g/mol for stoichiometric calculations.
  • Molecular formula C9H13N3O5.
  • Available in small research sizes suitable for assay development and biochemical assays.
  • Recommended storage: powder at -20°C or 4°C; in solution at -80°C or -20°C.

Catalog No. 50-002-83877 Supplier Medchemexpress LLC Supplier No. HYW5764405MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

α-Cytidine is a purine nucleoside analog used in biochemical and pharmacological research to investigate antitumor activity and nucleoside metabolism. It inhibits DNA synthesis and can induce apoptosis, making it relevant for studies of lymphoid malignancies and mechanistic assays.

  • Purity approximately 98.7% for reliable experimental results.
  • Molecular weight 243.22 g/mol for stoichiometric calculations.
  • Molecular formula C9H13N3O5.
  • Available in small research sizes suitable for assay development and biochemical assays.
  • Recommended storage: powder at -20°C or 4°C; in solution at -80°C or -20°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.