Learn More
Medchemexpress LLC Cytidine-13C | 201996-57-8 | 99.0% | 25 MG
SDP
Catalog No. 5000407408
Encompass_Preferred
Cytidine-13C is the 13C labeled Cytidine. Cytidine is a pyrimidine nucleoside and a component of RNA. It is also a precursor of uridine and controls neuronal-glial glutamate cycling, affecting cerebral phospholipid metabolism and catecholamine.
  • 13C labeled Cytidine
  • Pyrimidine nucleoside
  • Component of RNA
  • Precursor of uridine
  • Controls neuronal-glial glutamate cycling
  • Affects cerebral phospholipid metabolism
  • Affects catecholamine
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-07408 Supplier Medchemexpress LLC Supplier No. 25MGHYB0158S1
Cytidine-13C is the 13C labeled Cytidine. Cytidine is a pyrimidine nucleoside and a component of RNA. It is also a precursor of uridine and controls neuronal-glial glutamate cycling, affecting cerebral phospholipid metabolism and catecholamine.
  • 13C labeled Cytidine
  • Pyrimidine nucleoside
  • Component of RNA
  • Precursor of uridine
  • Controls neuronal-glial glutamate cycling
  • Affects cerebral phospholipid metabolism
  • Affects catecholamine
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS