Learn More
Medchemexpress LLC Cytidine-13C (Cytosine β-D-riboside-13C) | 201996-57-8 | 99.0% | 100 MG
SDP
Catalog No. 5000407243
Encompass_Preferred
Cytidine-13C is a 13C labeled pyrimidine nucleoside that acts as a component of RNA. It is a precursor of uridine and controls neuronal-glial glutamate cycling, affecting cerebral phospholipid metabolism and catecholamine. This compound is intended for research use only and is not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-07243 Supplier Medchemexpress LLC Supplier No. HYB0158S1100MG
Cytidine-13C is a 13C labeled pyrimidine nucleoside that acts as a component of RNA. It is a precursor of uridine and controls neuronal-glial glutamate cycling, affecting cerebral phospholipid metabolism and catecholamine. This compound is intended for research use only and is not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.