Learn More
Medchemexpress LLC Cytidine-13C9,15N3 | 202406-79-9 | 98.9% | 1 MG
SDP
Catalog No. 5000407197
Encompass_Preferred
Cytidine-13C9,15N3 is a 13C and 15N labeled Cytidine, a pyrimidine nucleoside that acts as a component of RNA and a precursor of uridine. It is involved in controlling neuronal-glial glutamate cycling, cerebral phospholipid metabolism, catecholamine synthesis, and mitochondrial function.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Component of RNA
  • Precursor of uridine
  • Involved in neuronal-glial glutamate cycling
  • Affects cerebral phospholipid metabolism
  • Influences catecholamine synthesis
  • Impacts mitochondrial function
Catalog No. 50-004-07197 Supplier Medchemexpress LLC Supplier No. HYB0158S51MG
Cytidine-13C9,15N3 is a 13C and 15N labeled Cytidine, a pyrimidine nucleoside that acts as a component of RNA and a precursor of uridine. It is involved in controlling neuronal-glial glutamate cycling, cerebral phospholipid metabolism, catecholamine synthesis, and mitochondrial function.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Component of RNA
  • Precursor of uridine
  • Involved in neuronal-glial glutamate cycling
  • Affects cerebral phospholipid metabolism
  • Influences catecholamine synthesis
  • Impacts mitochondrial function