Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cytidine-15N3 (cytosine β-D-riboside-15N3) | 65-46-3 | 99.3% | 246.20 | C9H13(15N)3O5 | 10 MG
SDP

Catalog No. 5000296824
Encompass_Preferred

Cytidine-15N3 is the 15N-labeled form of cytidine (cytosine β-D-riboside) used as a stable isotope tracer and internal standard in biochemical and analytical applications. It supports metabolic tracing and quantitative assays in research laboratories.

  • Stable isotope labeling for tracer and internal standard use.
  • High chemical purity (99.3%).
  • Suitable for mass spectrometry and isotope-dilution assays.
  • Provides molecular weight 246.20 and formula C9H13(15N)3O5 for accurate calculations.
  • Supplied as a 10 mg quantity for laboratory use.

Catalog No. 50-002-96824 Supplier Medchemexpress LLC Supplier No. HYB0158S610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cytidine-15N3 is the 15N-labeled form of cytidine (cytosine β-D-riboside) used as a stable isotope tracer and internal standard in biochemical and analytical applications. It supports metabolic tracing and quantitative assays in research laboratories.

  • Stable isotope labeling for tracer and internal standard use.
  • High chemical purity (99.3%).
  • Suitable for mass spectrometry and isotope-dilution assays.
  • Provides molecular weight 246.20 and formula C9H13(15N)3O5 for accurate calculations.
  • Supplied as a 10 mg quantity for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.