Learn More
Selleck Chemical LLC Cytidine S2053-50mg
SDP
Catalog No. 503108142
Encompass_Preferred
Cytidine is a nucleoside molecule that is formed when cytosine is attached to a ribose ring cytidine is a component of RNA
Catalog No. 50-310-8142 Supplier Selleck Chemical LLC Supplier No. S205350MG
Cytidine is a nucleoside molecule that is formed when cytosine is attached to a ribose ring cytidine is a component of RNA