Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cytidine, 2',3'-O-(1-methylethylidene)- | 362-42-5 | >=99.0% | C12H17N3O5 | 1 G
SDP

Catalog No. 5000353288
Encompass_Preferred

2',3'-O-Isopropylidenecytidine is a synthetic sugar compound intended for research use only.

  • Purity >=99.0%
  • Appearance: Solid, white to light yellow
  • Storage: Recommended storage at 4°C, protected from light; in solvent, -80°C for 6 months or -20°C for 1 month (protect from light)

Catalog No. 50-003-53288 Supplier Medchemexpress LLC Supplier No. HY1418841G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2',3'-O-Isopropylidenecytidine is a synthetic sugar compound intended for research use only.

  • Purity >=99.0%
  • Appearance: Solid, white to light yellow
  • Storage: Recommended storage at 4°C, protected from light; in solvent, -80°C for 6 months or -20°C for 1 month (protect from light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.