Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cytidine, 2',3'-O-(1-methylethylidene)- | 362-42-5 | ≥99.0% | C12H17N3O5 | 250 MG
SDP

Catalog No. 5000352558
Encompass_Preferred

2',3'-O-Isopropylidenecytidine is a synthetic sugar compound intended for research use only.

  • Purity of ≥99.0%
  • Molecular weight of 283.28
  • Chemical formula of C12H17N3O5
  • Appears as a white to light yellow solid
  • Recommended storage at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light

Catalog No. 50-003-52558 Supplier Medchemexpress LLC Supplier No. 250MGHY141884
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2',3'-O-Isopropylidenecytidine is a synthetic sugar compound intended for research use only.

  • Purity of ≥99.0%
  • Molecular weight of 283.28
  • Chemical formula of C12H17N3O5
  • Appears as a white to light yellow solid
  • Recommended storage at 4°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.