Learn More
Medchemexpress LLC HY-W008915 5mg Medchemexpress, Cytidine 5'-diphosphate (trisodium salt) CAS:34393-59-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW0089155MG
Encompass_Preferred
Medchemexpress, HY-W008915 5mg Cytidine 5'-diphosphate (trisodium salt) CAS:34393-59-4 Cytidine 5'-diphosphate trisodium salt (CDP) is produced by the transfer of phosphoryl group from ATP to cytidine monophosphate (CMP) catalyzed by uridine monophosphate kinase (UMPK). Cytidine 5′-diphosphate can be used to produce Cytidine triphosphate (CTP) for synthesis of DNA and RNA. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3263