Learn More
Medchemexpress LLC Cytidine 5'-diphosphate (trisodium salt) | 34393-59-4 | 98.0% | 469.12 | 10 MG
SDP
Catalog No. 5000451306
Encompass_Preferred
Cytidine 5'-diphosphate trisodium salt (CDP) is formed by the transfer of a phosphoryl group from ATP to cytidine monophosphate (CMP), catalyzed by uridine monophosphate kinase (UMPK). It is a key compound that can be utilized to produce Cytidine triphosphate (CTP) essential for DNA and RNA synthesis. This product also plays a role in the formation of CDP-ribitol, which is valuable for the diagnostics and research of CDP-l-ribitol pyrophosphorylase A (CRPPA) muscular dystrophy.
  • Involved in the enzymatic transfer of a phosphoryl group to cytidine monophosphate
  • Precursor for Cytidine triphosphate (CTP) in DNA and RNA synthesis
  • Useful for diagnostics and research related to CRPPA muscular dystrophy
Catalog No. 50-004-51306 Supplier Medchemexpress LLC Supplier No. HYW008915R10MG
Cytidine 5'-diphosphate trisodium salt (CDP) is formed by the transfer of a phosphoryl group from ATP to cytidine monophosphate (CMP), catalyzed by uridine monophosphate kinase (UMPK). It is a key compound that can be utilized to produce Cytidine triphosphate (CTP) essential for DNA and RNA synthesis. This product also plays a role in the formation of CDP-ribitol, which is valuable for the diagnostics and research of CDP-l-ribitol pyrophosphorylase A (CRPPA) muscular dystrophy.
  • Involved in the enzymatic transfer of a phosphoryl group to cytidine monophosphate
  • Precursor for Cytidine triphosphate (CTP) in DNA and RNA synthesis
  • Useful for diagnostics and research related to CRPPA muscular dystrophy