Learn More
Medchemexpress LLC Cytidine 5'-diphosphate (trisodium salt) | 34393-59-4 | 98.0% | 469.12 | 25 MG
SDP
Catalog No. 5000454014
Encompass_Preferred
Cytidine 5'-diphosphate trisodium salt (CDP) is generated through the transfer of a phosphoryl group from ATP to cytidine monophosphate (CMP), a reaction catalyzed by uridine monophosphate kinase (UMPK). It is utilized in the production of Cytidine triphosphate (CTP) for DNA and RNA synthesis.
  • Used for research purposes only.
  • CDP-ribitol can be used for the diagnostics and research of CDP-l-ribitol pyrophosphorylase A (CRPPA) muscular dystrophy.
  • Available in various quantities.
  • Can be stored at -20°C (protect from light, stored under nitrogen) or -80°C (in solvent) for different durations.
Catalog No. 50-004-54014 Supplier Medchemexpress LLC Supplier No. 25MGHYW008915R
Cytidine 5'-diphosphate trisodium salt (CDP) is generated through the transfer of a phosphoryl group from ATP to cytidine monophosphate (CMP), a reaction catalyzed by uridine monophosphate kinase (UMPK). It is utilized in the production of Cytidine triphosphate (CTP) for DNA and RNA synthesis.
  • Used for research purposes only.
  • CDP-ribitol can be used for the diagnostics and research of CDP-l-ribitol pyrophosphorylase A (CRPPA) muscular dystrophy.
  • Available in various quantities.
  • Can be stored at -20°C (protect from light, stored under nitrogen) or -80°C (in solvent) for different durations.