Learn More
Medchemexpress LLC Cytidine 5'-diphosphate (trisodium salt) | 34393-59-4 | 98.0% | 469.12 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW008915R5MG
Encompass_Preferred
Cytidine 5'-diphosphate trisodium salt (CDP) is formed by the transfer of a phosphoryl group from ATP to cytidine monophosphate (CMP), a reaction catalyzed by uridine monophosphate kinase (UMPK). It can be used to generate Cytidine triphosphate (CTP) for the synthesis of DNA and RNA.
  • Generated by phosphoryl group transfer from ATP to cytidine monophosphate (CMP)
  • Catalyzed by uridine monophosphate kinase (UMPK)
  • Can be used to produce cytidine triphosphate (CTP)
  • Suitable for synthesis of DNA and RNA
Catalog No. 50-003-66345