Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytidine-5'-diphosphate Trisodium Salt, ≥98%, MP Biomedicals™

Catalog No. ICN15075850 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15075850 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15075850 Supplier MP Biomedicals, Inc Supplier No. 0215075850
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34393-59-4
Molecular Formula C9H12N3Na3O11P2
Molecular Weight (g/mol) 469.12
MDL Number MFCD00065201
InChI Key JENUKVZGXGULDX-LLWADOMFSA-K
Synonym cytidine 5'-diphosphate trisodium salt
PubChem CID 131674171
IUPAC Name trisodium ({[(2R,3S,4R,5R)-5-(4-amino-2-oxo-1,2-dihydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
SMILES [Na+].[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])([O-])=O)[C@@H](O)[C@H]1O

Specifications

Assay Percent Range ≥98%
Quantity 50 mg
Formula Weight 469.12
Physical Form Crystalline Powder
Chemical Name or Material Cytidine-5'-diphosphate Trisodium Salt
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.