Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cytidine 5'-diphosphoethanolamine | 3036-18-8 | 99.3% | 446.24 g/mol | C11H20N4O11P2 | 5 MG
SDP

Catalog No. 5000221499
Encompass_Preferred

Cytidine 5′-diphosphoethanolamine is a nucleotide-derived intermediate used in the biosynthetic and chemical synthesis of phosphatidylethanolamine. Supplied as a high-purity research reagent, it is used in studies of lipid metabolism and cholinergic function.

  • Intermediate in phosphatidylethanolamine synthesis.
  • Reported stimulant of acetylcholine synthesis.
  • High purity (99.3%).
  • Molecular weight 446.24 g/mol; formula C11H20N4O11P2.
  • Supplied as a 5 mg powder for research use.
  • Storage: powder at -20°C; in solution -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-21499 Supplier Medchemexpress LLC Supplier No. HY1457805MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cytidine 5′-diphosphoethanolamine is a nucleotide-derived intermediate used in the biosynthetic and chemical synthesis of phosphatidylethanolamine. Supplied as a high-purity research reagent, it is used in studies of lipid metabolism and cholinergic function.

  • Intermediate in phosphatidylethanolamine synthesis.
  • Reported stimulant of acetylcholine synthesis.
  • High purity (99.3%).
  • Molecular weight 446.24 g/mol; formula C11H20N4O11P2.
  • Supplied as a 5 mg powder for research use.
  • Storage: powder at -20°C; in solution -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.