Learn More
Medchemexpress LLC Cytidine 5'-diphosphoethanolamine | 3036-18-8 | 99.3% | C11H20N4O11P2 | 10MG
SDP
Catalog No. 5000261562
Encompass_Preferred
Cytidine 5′-diphosphoethanolamine is a nucleotide derivative used as an intermediate in the biosynthetic and synthetic pathways for phosphatidylethanolamine. It is employed as a research reagent and has been reported to stimulate acetylcholine (Ach) synthesis, making it useful in studies of phospholipid metabolism and cholinergic signaling.
  • Intermediate in phosphatidylethanolamine synthesis.
  • Reported stimulant of acetylcholine synthesis.
  • Suitable for research on phospholipid metabolism and cholinergic systems.
  • High reported purity for analytical and preparative applications.
  • Available in small milligram pack sizes for laboratory use.
Catalog No. 50-002-61562 Supplier Medchemexpress LLC Supplier No. HY14578010MG
Cytidine 5′-diphosphoethanolamine is a nucleotide derivative used as an intermediate in the biosynthetic and synthetic pathways for phosphatidylethanolamine. It is employed as a research reagent and has been reported to stimulate acetylcholine (Ach) synthesis, making it useful in studies of phospholipid metabolism and cholinergic signaling.
  • Intermediate in phosphatidylethanolamine synthesis.
  • Reported stimulant of acetylcholine synthesis.
  • Suitable for research on phospholipid metabolism and cholinergic systems.
  • High reported purity for analytical and preparative applications.
  • Available in small milligram pack sizes for laboratory use.