Learn More
Medchemexpress LLC Cytidine 5'-monophosphate | 63-37-6 | 99.9% | 323.20 | 25MG
SDP
Catalog No. 5000453263
Encompass_Preferred
Cytidine 5'-monophosphate (5'-Cytidylic acid) is a nucleotide which is used as a monomer in RNA. It consists of the nucleobase cytosine, the pentose sugar ribose, and a phosphate group. This product is intended for research and analytical applications and can serve as a drug intermediate for the synthesis of nucleotide derivatives, used in food and pharmaceutical industries.
  • Used as a monomer in RNA
  • Composed of cytosine, ribose, and a phosphate group
  • Functions as an analytical standard
  • Intended for research and analytical applications
  • Serves as a drug intermediate for nucleotide derivative synthesis
  • Applicable in food and pharmaceutical industries
Catalog No. 50-004-53263 Supplier Medchemexpress LLC Supplier No. 25MGHYW009162R
Cytidine 5'-monophosphate (5'-Cytidylic acid) is a nucleotide which is used as a monomer in RNA. It consists of the nucleobase cytosine, the pentose sugar ribose, and a phosphate group. This product is intended for research and analytical applications and can serve as a drug intermediate for the synthesis of nucleotide derivatives, used in food and pharmaceutical industries.
  • Used as a monomer in RNA
  • Composed of cytosine, ribose, and a phosphate group
  • Functions as an analytical standard
  • Intended for research and analytical applications
  • Serves as a drug intermediate for nucleotide derivative synthesis
  • Applicable in food and pharmaceutical industries