Learn More
Abcam Cytidine-5'-monophosphate (CMP), Nucleoside monophosphate, 100MG
Catalog No. 502704837
Encompass_Preferred
MW 323.2 g/mol, Purity >95%. Nucleoside monophosphate used as a substrate of uridine monophosphate (UMP)/cytidine monophosphate (CMP) kinase (EC 2.7.4.4) to form CDP which upon phosphorylation to CTP supports DNA and RNA biosynthesis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4837 Supplier Abcam Supplier No. AB146215100MG
MW 323.2 g/mol, Purity >95%. Nucleoside monophosphate used as a substrate of uridine monophosphate (UMP)/cytidine monophosphate (CMP) kinase (EC 2.7.4.4) to form CDP which upon phosphorylation to CTP supports DNA and RNA biosynthesis.
The product is subject to the following: Abcam Restricted Use Statement