Learn More
Medchemexpress LLC Cytidine 5'-monophosphate-d12 dilithium | 347.14 | 1 MG
SDP
Catalog No. 5000453372
Encompass_Preferred
Cytidine 5′-monophosphate-d12 dilithium is a deuterium-labeled variant of Cytidine 5'-monophosphate, a nucleotide serving as a monomer in RNA. It comprises the nucleobase cytosine, the pentose sugar ribose, and a phosphate group. This deuterium labeling is utilized as a tracer for quantitation in drug development and can influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled nucleotide.
  • Used as a monomer in RNA.
  • Comprises cytosine, ribose, and a phosphate group.
  • Functions as a tracer for drug quantitation.
  • Influences pharmacokinetic and metabolic profiles.
Catalog No. 50-004-53372 Supplier Medchemexpress LLC Supplier No. HYW009162S21MG
Cytidine 5′-monophosphate-d12 dilithium is a deuterium-labeled variant of Cytidine 5'-monophosphate, a nucleotide serving as a monomer in RNA. It comprises the nucleobase cytosine, the pentose sugar ribose, and a phosphate group. This deuterium labeling is utilized as a tracer for quantitation in drug development and can influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled nucleotide.
  • Used as a monomer in RNA.
  • Comprises cytosine, ribose, and a phosphate group.
  • Functions as a tracer for drug quantitation.
  • Influences pharmacokinetic and metabolic profiles.