Learn More
Medchemexpress LLC Cytidine 5'-monophosphate (Standard) | 63-37-6 | 97% | 323.20 | 100 MG
SDP
Catalog No. 5000366861
Encompass_Preferred
Cytidine 5'-monophosphate (Standard), also known as 5'-Cytidylic acid (Standard) or 5'-CMP (Standard), is an analytical standard used for research and analytical applications. It is a nucleotide that serves as a monomer in RNA and is composed of the nucleobase cytosine, the pentose sugar ribose, and a phosphate group.
  • Analytical standard supplied assay for reference.
  • Commonly used in qualitative, quantitative, and methodological research experiments.
  • Suitable for use in HPLC, GC, and MS applications.
Catalog No. 50-003-66861 Supplier Medchemexpress LLC Supplier No. HYW009162R100MG
Cytidine 5'-monophosphate (Standard), also known as 5'-Cytidylic acid (Standard) or 5'-CMP (Standard), is an analytical standard used for research and analytical applications. It is a nucleotide that serves as a monomer in RNA and is composed of the nucleobase cytosine, the pentose sugar ribose, and a phosphate group.
  • Analytical standard supplied assay for reference.
  • Commonly used in qualitative, quantitative, and methodological research experiments.
  • Suitable for use in HPLC, GC, and MS applications.