Learn More
Medchemexpress LLC Cytidine-5'-triphosphate-15N3 dilithium | 65-47-4 | 97.0% | 498.00 | 1 MG
SDP
Catalog No. 5000438583
Encompass_Preferred
Cytidine-5'-triphosphate-15N3 dilithium (5'-CTP-15N3) is 15N labeled Cytidine-5'-triphosphate. It is a nucleoside triphosphate that serves as a building block for nucleotides and nucleic acids, and lipid biosynthesis. Cytidine triphosphate synthase can catalyze the formation of cytidine 5′-triphosphate from uridine 5′-triphosphate (UTP). This compound is an essential biomolecule in the de novo pyrimidine biosynthetic pathway in *T. gondii*.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-38583 Supplier Medchemexpress LLC Supplier No. HY125818S61MG
Cytidine-5'-triphosphate-15N3 dilithium (5'-CTP-15N3) is 15N labeled Cytidine-5'-triphosphate. It is a nucleoside triphosphate that serves as a building block for nucleotides and nucleic acids, and lipid biosynthesis. Cytidine triphosphate synthase can catalyze the formation of cytidine 5′-triphosphate from uridine 5′-triphosphate (UTP). This compound is an essential biomolecule in the de novo pyrimidine biosynthetic pathway in *T. gondii*.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.