Learn More
Selleck Chemical LLC Cytidine 5-triphosphate
SDP
Catalog No. 5000000076
Encompass_Preferred
Cytidine 5 -triphosphate is a pyrimidine nucleoside triphosphate that is involved in a variety of biochemical reactions It is used in the synthesis of RNA by RNA polymerases
Catalog No. 50-000-00076 Supplier Selleck Chemical LLC Supplier No. S607725MG
Cytidine 5 -triphosphate is a pyrimidine nucleoside triphosphate that is involved in a variety of biochemical reactions It is used in the synthesis of RNA by RNA polymerases