Learn More
Apexbio Technology LLC Cytidine 5-triphosphate 65-47-4 100mg
SDP
Supplier:  Apexbio Technology LLC B7880100
Encompass_Preferred
Cytidine 5 -triphosphate (CTP CAS 65-47-4) is a pyrimidine nucleoside triphosphate that serves as a substrate in various biochemical pathways notably as a precursor for RNA synthesis and as a cofactor in phospholipid and glycoprotein biosynthesis CTP participates in the rate-limiting conversion of phosphocholine to CDP-choline catalyzed by CTP phosphocholine cytidylyltransferase a critical step in phosphatidylcholine production This process is implicated in membrane biogenesis and the provision of substrates for cell growth Additionally CTP is utilized by N-acylneuraminate cytidylyltransferase to form CMP-Neu5Ac an essential intermediate in sialylation CTP is widely employed in studies of nucleotide metabolism membrane synthesis and glycosylation pathways
Catalog No. 50-000-92287