Learn More
Medchemexpress LLC Cytidine 5'-triphosphate | 65-47-4 | 98.0% | 483.16 | C9H16N3O14P3 | 1G
SDP
Catalog No. 5000239101
Encompass_Preferred
Cytidine 5'-triphosphate (CTP) is a ribonucleoside triphosphate used as a nucleotide building block for RNA synthesis and as a co-substrate in lipid biosynthesis. Supplied as a research-grade white powder, it is used as a substrate in enzymatic assays, in vitro transcription, and studies of pyrimidine metabolism. The material has molecular weight 483.16 g/mol and chemical formula C9H16N3O14P3.
Catalog No. 50-002-39101 Supplier Medchemexpress LLC Supplier No. HY1258181G
Cytidine 5'-triphosphate (CTP) is a ribonucleoside triphosphate used as a nucleotide building block for RNA synthesis and as a co-substrate in lipid biosynthesis. Supplied as a research-grade white powder, it is used as a substrate in enzymatic assays, in vitro transcription, and studies of pyrimidine metabolism. The material has molecular weight 483.16 g/mol and chemical formula C9H16N3O14P3.